Thifluzamide ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 2′,6′-Dibromo-2-methyl-4′-trifluoromethoxy-4-trifluoromethyl-1,3-thiazole-5-carboxanilide

CDX-T0118-M010 CDX-T0118-M050
102991-128EA 105.23 USD
102991-128 102991-130
Thifluzamide ≥97% (by HPLC)
Thifluzamide

Thiazole fungicide used in agrochemistry. Succinate dehydrogenase inhibitor (SDHI) which interferes with succinate ubiquinone reductase in the mitochondrial electron transport chain of fungi.Compound can be used as analytical reference material.

Formula: C₁₃H₆Br₂F₆N₂O₂S
MW: 528.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03428071
CAS Number: 130000-40-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (HPLC)
Appearance/Color White to beige or faint brown powder.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR