2,4,6-Triiodobenzoic acid ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-T0095-M100 CDX-T0095-M500
102991-112EA 175.85 USD
102991-112 102991-114
2,4,6-Triiodobenzoic acid ≥98% (by HPLC)
2,4,6-Triiodobenzoic acid

Useful as contrast media agent in radiology.

Formula: C₇H₃I₃O₂
Storage Temperature: Freezer
CAS Number: 2012-31-9

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in chloroform or toluene.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR