1,2,3-Tris(diethylamino)cyclopropenylium dicyanamide ≥97%
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-T0089-G001
CDX-T0089-G005
102991-104EA
78.92
USD
102991-104
102991-106
1,2,3-Tris(diethylamino)cyclopropenylium dicyanamide ≥97%
1,2,3-Tris(diethylamino)cyclopropenylium dicyanamide
Ionic liquid. The properties of these ionic liquids include almost zero vapour pressure, low flammability and ease of recycling. They are frequently considered as potential green alternatives to classical organic solvents. Additionally, most classes can be readily tuned to provide unique solvent environments in which to carry out reactions, extractions and other processes. There are a large number of applications.
Formula:
C₁₇H₃₀N₆ Storage Temperature: Freezer |
CAS Number:
1415962-26-3 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | Yellow to orange viscous liquid. |
Solubility | Soluble in chloroform or water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...