4-(Trifluoroacetyl)benzoyl chloride ≥97% (by GC)

Supplier: ADIPOGEN CORP MS
Danger

CDX-T0082-G001 CDX-T0082-G005 CDX-T0082-G050
102991-090EA 84.46 USD
102991-090 102991-092 102991-094
4-(Trifluoroacetyl)benzoyl chloride ≥97% (by GC)
4-(Trifluoroacetyl)benzoyl chloride

Building block for synthesis.

Formula: C₉H₄ClF₃O₂
Storage Temperature: Refrigerator
CAS Number: 58808-60-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color Colourless to slightly yellow liquid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR