1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]benzindolium inner salt ≥90%

Supplier: ADIPOGEN CORP MS

Synonyms: 1,1,2-Trimethyl-3-(4-sulphobutyl)-1h-benz[e]indoliumhydroxide,innersalt

CDX-T0061-M500 CDX-T0061-GG25
102991-082EA 178.62 USD
102991-082 102991-084
1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]benzindolium inner salt ≥90%
1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]benzindolium inner salt

Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.

Formula: C₁₉H₂₃NO₃S
Storage Temperature: Freezer
MDL Number: MFCD02690163
CAS Number: 63149-24-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity ~90% (Dye Content) (UV/Vis)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR