2,3,3-Trimethyl-1-(4-sulfobutyl)-indolium, inner salt ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
4-(2,3,3-Trimethyl-3H-indol-1-ium-1-yl)butane-1-sulphonate
CDX-T0042-M100
CDX-T0042-M500
CDX-T0042-G005
102991-068EA
95.54
USD
102991-068
102991-070
102991-072
2,3,3-Trimethyl-1-(4-sulfobutyl)-indolium, inner salt ≥98% (by HPLC)
2,3,3-Trimethyl-1-(4-sulfobutyl)indolium, inner salt
Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.
Formula:
C₁₅H₂₁NO₃S MW: 295.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD08166699 CAS Number: 54136-26-4 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...