Tetramethylrhodamine isothiocyanate Isomer R (6-TRITC isomer R) ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: Tetramethylrhodamine-6-isothiocyanate

CDX-T0035-M010 CDX-T0035-M100
102991-064EA 202.15 USD
102991-064 102991-066
Tetramethylrhodamine isothiocyanate Isomer R (6-TRITC isomer R) ≥95%
Tetramethylrhodamine isothiocyanate Isomer R (6-TRITC isomer R)

Useful in the fluorescent labeling of proteins. Labeling efficiency with bovine albumin: > 50%.

Formula: C₂₅H₂₁N₃O₃S
MW: 443.52 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063517
CAS Number: 80724-20-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >95% (Elemental Analysis)
Appearance/Color Deep violet powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR