Tetramethylrhodamine-6-maleimide ≥97%
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-T0030-M002
CDX-T0030-M025
102991-056EA
263.08
USD
102991-056
102991-058
Tetramethylrhodamine-6-maleimide ≥97%
Tetramethylrhodamine-6-maleimide
Fluorescent reagent with high selectivity.
Formula:
C₂₈H₂₃N₃O₅ MW: 481.5 g/mol Storage Temperature: Refrigerator |
CAS Number:
174568-68-4 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in methanol, DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...