Tetramethylrhodamine-5-maleimide ≥85%

Supplier: ADIPOGEN CORP MS

Synonyms: TMR-5 Malemide, 5-TAMRA Maleimide

CDX-T0029-M001 CDX-T0029-M005 CDX-T0029-M025
102991-050EA 141.23 USD
102991-050 102991-052 102991-054
Tetramethylrhodamine-5-maleimide ≥85%
Tetramethylrhodamine-5-maleimide

Labeling reagent for protein thiols for cytofluorescence.

Formula: C₂₈H₂₃N₃O₅
MW: 481.5 g/mol
Storage Temperature: Freezer
CAS Number: 174568-67-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 85% (HPLC)
Appearance/Color Pink solid.
Solubility Soluble in methanol, DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR