(+)-Spartein surrogate 96
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
(1R,2S,9S)-(+)-11-Methyl-7,11-diazatricyclo[7.3.1.02,7]tridecane
CDX-S0045-M100
CDX-S0045-G001
102991-010EA
279.69
USD
102991-010
102991-012
(+)-Spartein surrogate 96
(+)-Spartein surrogate
Chiral reagent.
Formula:
C₁₂H₂₂N₂ MW: 194.32 g/mol Storage Temperature: Freezer |
CAS Number:
475301-86-1 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Viscous oil. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...