SNARF-DE ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Chromoionophore XIII
CDX-S0037-M010
CDX-S0037-M050
102991-002EA
88.62
USD
102991-002
102991-004
SNARF-DE ≥95%
SNARF-DE
Lipophilic fluorescent pH indicator used to assay biogenic amines with optodes. The deprotonated version enables excitation with red light (LED) (Ex/Em: 605/680nm) versus the acidic form (Ex/Em: 520/600nm). The ester functionality renders the indicator molecule lipophilic and water-insoluble but also prevents lactonization of the dye in an apolar environment.
Formula:
C₃₈H₄₄NO₄·ClO₄ MW: 678.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00799316 CAS Number: 135656-96-1 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Deep red powder. |
Solubility | Soluble in dichloromethane. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...