Sorbic chloride ≥90%
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
Sorbyl chloride
Reactant for: (1) preparation of unsaturated hydroxymethylene diphosphonic acid derivatives by reaction of trimethylsilyl phosphites, (2) asymmetrical synthesis of the isoindolinone subunit of the macrocyclic poylketide natural product muironolide via stereoselective intramolecular Diels-Alder cycloaddition, (3) preparation of emollient, humectant, and fluorescent alpha,beta-unsaturated thiol esters for long-acting skin applications, (4) stereoselective synthesis of enediol carbonates for use in the asymmetrical synthesis of alpha-acyloxyketones, (5) acylation of pyrazoles.
Formula:
C₆H₇ClO MW: 130.57 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00045227 CAS Number: 2614-88-2 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | Colourless to yellow liquid. |
Solubility | Soluble in water or dichloromethane. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...