Sodium tetra(p-tolyl)borate ≥99%

Supplier: ADIPOGEN CORP MS
Danger

CDX-S0032-M500 CDX-S0032-G010
102990-994EA 192.46 USD
102990-994 102990-996
Sodium tetra(p-tolyl)borate ≥99%
Sodium tetra(p-tolyl)borate

Analytical/Chromatography reagent. Ion sensor compound. Additive for ISE.

Formula: C₂₈H₂₈BNa
Storage Temperature: Freezer
CAS Number: 15738-23-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 99% (Titration)
Appearance/Color Solid.
Solubility Soluble in dichloromethane.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR