N-(3-Succinimidyloxy-carbonyl-phenyl)-methyl-4-(2-(6-(3,4-dihydro-2H-1-benzopyranyl))-5-oxazolyl)-pyridinium bromide ≥90% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

CDX-S0031-M005 CDX-S0031-M050 CDX-S0031-M250
102990-988EA 96.92 USD
102990-988 102990-990 102990-992
N-(3-Succinimidyloxy-carbonyl-phenyl)-methyl-4-(2-(6-(3,4-dihydro-2H-1-benzopyranyl))-5-oxazolyl)-pyridinium bromide ≥90% (by NMR)
N-(3-Succinimidyloxycarbonylphenyl)methyl-4-(2-(6-(3,4-dihydro-2H-1-benzopyranyl))-5-oxazolyl)pyridinium bromide

New class of highly photostable, water soluble fluorescent labels with large Stoke′s shift, high QY in aqueous media and pH tolerance.

Formula: C₂₉H₂₄BrN₃O₆
MW: 590.42 g/mol
Storage Temperature: Freezer
CAS Number: 155863-03-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (NMR)
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR