[Ru(bpy)2(5-chloroacetamido-1,10-phenanthroline)](PF6)2 ≥95%

Supplier: ADIPOGEN CORP MS
Warning

CDX-R0028-M010 CDX-R0028-M100
102990-944EA 245.08 USD
102990-944 102990-946
[Ru(bpy)2(5-chloroacetamido-1,10-phenanthroline)](PF6)2 ≥95%
[Ru(bpy)2(5-chloroacetamido-1,10-phenanthroline)](PF6)2

Reactive probe containing s fluorescent metal-ligand complex for the determination of nucleotide base sequences.

Formula: C₃₄H₂₆F₁₂ClN₇OP₂Ru
Storage Temperature: Freezer
CAS Number: 204273-42-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in dichloromethane, toluene or acetonitrile.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR