Rhodamine 6G bis(aminoethyl)amine amide bis (trifluoroacetate) ≥95% (by HPCE)

Supplier: ADIPOGEN CORP MS
Warning

CDX-R0025-M050 CDX-R0025-M250
102990-940EA 211.85 USD
102990-940 102990-942
Rhodamine 6G bis(aminoethyl)amine amide bis (trifluoroacetate) ≥95% (by HPCE)
Rhodamine 6G bis(aminoethyl)amine amide bis(trifluoroacetate)

Fluorescent labelling.

Formula: C₃₀H₃₇N₅O₂·C₄H₂F₆O₄
Storage Temperature: Freezer
CAS Number: 1173097-37-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR