Rhodamine 6G bis(oxyethylamino)ethane amide bis (trifluoroacetate) ≥95% (by HPCE)

Supplier: ADIPOGEN CORP MS

CDX-R0023-M050 CDX-R0023-M250
102990-932EA 211.85 USD
102990-932 102990-934
Rhodamine 6G bis(oxyethylamino)ethane amide bis (trifluoroacetate) ≥95% (by HPCE)
Rhodamine 6G bis(oxyethylamino)ethane amide bis(trifluoroacetate)

Fluorescent labelling.

Formula: C₃₂H₄₀N₄O₄·C₄H₂F₆O₄
Storage Temperature: Freezer
CAS Number: 1173097-69-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Off-white solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR