Rhodamine 6G ethylenediamine amide bis (trifluoroacetate) ≥95% (by HPCE)

Supplier: ADIPOGEN CORP MS
Warning

CDX-R0022-M010 CDX-R0022-M050
102990-928EA 123.23 USD
102990-928 102990-930
Rhodamine 6G ethylenediamine amide bis (trifluoroacetate) ≥95% (by HPCE)
Rhodamine 6G ethylenediamine amide bis(trifluoroacetate)

Fluorescent labelling.

Formula: C₂₈H₃₂N₄O₂·C₄H₂F₆O₄
MW: 684.63 g/mol
Storage Temperature: Freezer
CAS Number: 591742-74-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Light pink solid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR