Bis(2,2′-bipyridine)-(5-isothiocyanato-phenanthroline)ruthenium bis(hexafluorophosphate) ≥95% (by NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: Ru(bpy)2(phen-ITC)2+

CDX-R0018-M001 CDX-R0018-M005 CDX-R0018-M025
102990-918EA 84.46 USD
102990-918 102990-920 102990-922
Bis(2,2′-bipyridine)-(5-isothiocyanato-phenanthroline)ruthenium bis(hexafluorophosphate) ≥95% (by NMR)
Bis(2,2′-bipyridine)(5-isothiocyanatophenanthroline)ruthenium bis(hexafluorophosphate)

Reactive probe containing fluorescent metal-ligand complex for determination of nucleotide base sequences.

Formula: C₃₃H₂₃F₁₂N₇P₂RuS
MW: 940.65 g/mol
Storage Temperature: Freezer
CAS Number: 288399-07-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (NMR)
Appearance/Color Orange powder.
Solubility Soluble in dichloromethane, toluene or acetonitrile.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR