Quinacrine mustard dihydrochloride ≥95%
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
2-Methoxy-6-chloro-9-[4-bis(2-chloroethyl)amino-1-methylbutyl amino]acridine dihydrochloride
CDX-Q0011-M010
CDX-Q0011-M025
CDX-Q0011-M100
102990-890EA
123.23
USD
102990-890
102990-892
102990-894
Quinacrine mustard dihydrochloride ≥95%
Quinacrine mustard dihydrochloride
Fluorescent probe for labeling chromosomal DNA. Also used in measurement of sequence specificity with Taq DNA polymerase.
Formula:
C₂₃H₂₈Cl₃N₃O·2HCl MW: 541.8 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00043254 CAS Number: 4213-45-0 UN: 2811 ADR: 6.1 III |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Orange to dark yellow powder. |
Solubility | Soluble in water or chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...