2-Propyl-2-pentenoic acid ≥98% (by GC)

Supplier: ADIPOGEN CORP MS
Danger

CDX-P0155-M025 CDX-P0155-M050
102990-880EA 360 USD
102990-880 102990-882
2-Propyl-2-pentenoic acid ≥98% (by GC)
2-Propyl-2-pentenoic acid

A major beta-oxidation metabolite of valproic acid (VPA). Exhibits anticonvulsive potency similar to VPA and may be at least partially responsible for its neurological activity. Compound can be used as analytical reference material.

Formula: C₈H₁₄O₂
Storage Temperature: Freezer
MDL Number: MFCD00801107
CAS Number: 60218-41-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (GC)
Appearance/Color White crystalline powder.
Solubility Soluble in chloroform, ether or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR