N-Phenyl-benzimidoyl chloride ≥95% (by NMR)

Supplier: ADIPOGEN CORP MS
Danger

CDX-P0120-G001 CDX-P0120-GG25 CDX-P0120-G025
102990-844EA 113.54 USD
102990-844 102990-846 102990-848
N-Phenyl-benzimidoyl chloride ≥95% (by NMR)
N-Phenyl-benzimidoyl chloride

Building block for synthesis.

Formula: C₁₃H₁₀ClN
Storage Temperature: Refrigerator
CAS Number: 4903-36-0
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (NMR)
Appearance/Color Solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR