N-Phenylhydroxylamine ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Danger

CDX-P0103-G001 CDX-P0103-G005 CDX-P0103-G025
102990-822EA 113.54 USD
102990-822 102990-824 102990-826
N-Phenylhydroxylamine ≥98% (by HPLC)
N-Phenylhydroxylamine

Building block for synthesis.

Formula: C₆H₇NO
MW: 109.13 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00045718
CAS Number: 100-65-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Fluffy white solid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR