Propionaldehyde oxime ≥96% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Propanal oxime

CDX-P0059-M500 CDX-P0059-GG25 CDX-P0059-G010
102990-808EA 78.92 USD
102990-808 102990-810 102990-812
Propionaldehyde oxime ≥96% (by HPLC)
Propionaldehyde oxime

Building block for synthesis.

Formula: C₃H₇NO
MW: 73.09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00516618
CAS Number: 627-39-4

Order Now

Specification Test Results

Source/Host Synthetic
Purity 96% (HPLC)
Appearance/Color Colourless liquid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR