Oxonol V ≥95% (by UV method)

Supplier: ADIPOGEN CORP MS
Warning

CDX-P0019-M050 CDX-P0019-M500
102990-780EA 170.31 USD
102990-780 102990-782
Oxonol V ≥95% (by UV method)
Oxonol V

Potential sensitive probe. Determination of plasma membrane potential of neutrophils generated by the Na+ pump.

Formula: C₂₃H₁₆N₂O₄
Storage Temperature: Freezer
CAS Number: 61389-30-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (UV)
Appearance/Color Dark blue solid.
Solubility Soluble in methanol, ethanol, DMSO, DMF or acetonitrile.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR