Poly(D-lactide), M.W. 90000-124000 g/mol, (max. 0.5% H₂O)
Supplier: ADIPOGEN CORP MS
CDX-P0018-M100
CDX-P0018-M250
CDX-P0018-GG25
102990-774EA
210.46
USD
102990-774
102990-776
102990-778
Poly(D-lactide), M.W. 90000-124000 g/mol, (max. 0.5% H₂O)
Poly(D-lactide)
Poly(D-lactide) (PDLA), a polymer of a stereospecific cyclic di-ester of lactic acid, is used in biomaterial research for the development of devices such as therapeutic drug delivery vessels. Poly(D-lactide) is used for the preparation of microparticles and resorbable polylactide scaffolds.
Formula:
(C₃H₄O₂)n Storage Temperature: Refrigerator |
CAS Number:
106989-11-1 |
Specification Test Results
Source/Host | Synthetic. |
Purity | <0.5% (water) |
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...