Poly(L-lactide), M.W. 66,000-107,000 g/mol 2.0 dl/g, (max. 0.5% H₂O)

Supplier: ADIPOGEN CORP MS

Synonyms: PLA

CDX-P0016-G010 CDX-P0016-G050
102990-770EA 192.46 USD
102990-770 102990-772
Poly(L-lactide), M.W. 66,000-107,000 g/mol 2.0 dl/g, (max. 0.5% H₂O)
Poly(L-lactide)

Polylactides (PLA) used in material sciences. Polymer. Lactide and Glycolide Polymers.


  • 2.0 dl/g

Formula: (C₃H₄O₂)n
CAS Number: 26161-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR