Poly(L-lactide), M.W. 59,000-101,000 g/mol 1.0 dl/g, (max. 0.5% H₂O)
Supplier: ADIPOGEN CORP MS
Synonyms:
PLA
CDX-P0013-G010
CDX-P0013-G050
102990-762EA
193.85
USD
102990-762
102990-764
Poly(L-lactide), M.W. 59,000-101,000 g/mol 1.0 dl/g, (max. 0.5% H₂O)
Poly(L-lactide)
Polylactides (PLA) used in material sciences. Lactide and Glycolide Polymers.
- 1.0 dl/g
Formula:
(C₃H₄O₂)n Storage Temperature: Refrigerator |
CAS Number:
26161-42-2 |
Specification Test Results
Source/Host | Synthetic. |
Purity | <0.5% (water) |
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...