Physil chloride ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-P0009-M050 CDX-P0009-M250
102990-758EA 81.69 USD
102990-758 102990-760
Physil chloride ≥98% (by HPLC)
Physil chloride

It is known that phenol and p-cresol are promoters of skin tumors and also urinary excretion of phenol and p-cresol is increased by exposure to benzene, which has hemopoietic toxicity, and toluene, respectively. Therefore, the determination of phenol and p-cresol is important in clinical chemistry and toxicology. Because phenol and p-cresol are present in urine as sulfate or glucuronide conjugates, they are hydrolyzed enzymatically or with acid, followed by distillation or extraction with organic solvent prior to measurement by GC or HPLC. However, these GC and HPLC methods require relatively large sample volumes (more than 1 ml) because of their insufficient sensitivities for measuring small amounts of phenol and p-cresol. The application of fluorescent labeling techniques to HPLC result in a sensitive and selective method. Phthalimidylbenzenesulfonyl chlorides habe also been successfully used for high sensitivity labelling of amino acids prior to HPLC.

Formula: C₁₄H₁₀ClNO₃S
Storage Temperature: Refrigerator
CAS Number: 114341-14-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Solid.
Solubility Soluble in acetone.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR