12-(1-Pyrenyl) dodecanoic acid ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 12-(Pyren-1-yl)dodecanoic acid

CDX-P0006-M005 CDX-P0006-M050
102990-754EA 139.85 USD
102990-754 102990-756
12-(1-Pyrenyl) dodecanoic acid ≥98% (by HPLC)
12-(1-Pyrenyl)dodecanoic acid

Anionic membrane probe used for studies on the uptake of fluorescent fatty acids into cultured cells and oxygen sensing.

Formula: C₂₈H₃₂O₂
MW: 400.55 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00080927
CAS Number: 69168-45-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Solid.
Solubility Soluble in acetone. Slightly soluble in methanol (0.5mg/ml) or DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR