N-(3-Oxooctanoyl)-L-homoserine lactone 97

Supplier: ADIPOGEN CORP MS

Synonyms: N-3-Oxo-octanoyl-L-homoserine lactone, (S)-4-Oxo-N-(2-oxotetrahydrofuran-3-yl)nonanamide, 3-oxo-C8-HSL

CDX-O0058-M010 CDX-O0058-M100
102990-734EA 88.62 USD
102990-734 102990-736
N-(3-Oxooctanoyl)-L-homoserine lactone 97
N-(3-Oxooctanoyl)-L-homoserine lactone

N-(3-Oxooctanoyl)-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. This compound stimulates the tra gene expression. It is an autoinducer and potent antagonist. Promotes the expression of the transcriptional activator (and LuxR homolog) TraR, in the gram-negative bacterium A. tumefaciens.

Formula: C₁₂H₁₉NO₄
MW: 241.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08277025
CAS Number: 147795-39-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color White solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR