N-(3-Oxotetradecanoyl)-DL-homoserine lactone ≥97% (by GC)
Supplier: ADIPOGEN CORP MS
Synonyms:
3-oxo-C14-HSL
N-(3-Oxotetradecanoyl)-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. It appears later than shorter acyl chain AHLs in developing biofilms and like other long chain 3-oxo-AHLs, stimulates the production of putisolvin, which in turn inhibits biofilm formation.
Formula:
C₁₈H₃₁NO₄ MW: 325.4 g/mol Storage Temperature: Freezer |
CAS Number:
503610-29-5 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...