N-(3-Oxooctanoyl)-DL-homoserine lactone ≥97% (by GC)

Supplier: ADIPOGEN CORP MS

Synonyms: N-(3-Oxohexanoyl)-DL-homoserine lactone, 3-oxo-C8-HSL

CDX-O0052-M025 CDX-O0052-M100
102990-714EA 105.23 USD
102990-714 102990-716
N-(3-Oxooctanoyl)-DL-homoserine lactone ≥97% (by GC)
N-(3-Oxooctanoyl)-DL-homoserine lactone

N-(3-Oxooctanoyl)-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. This compound stimulates the tra gene expression. It is an autoinducer and potent antagonist.

Formula: C₁₂H₁₉NO₄
MW: 241.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12912431
CAS Number: 106983-27-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color White solid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR