N-(3-Oxobutanoyl)-DL-homoserine lactone ≥97% (by GC)

Supplier: ADIPOGEN CORP MS

Synonyms: 3-oxo-C4-HSL

CDX-O0050-M010 CDX-O0050-M050
102990-706EA 88.62 USD
102990-706 102990-708
N-(3-Oxobutanoyl)-DL-homoserine lactone ≥97% (by GC)
N-(3-Oxobutanoyl)-DL-homoserine lactone

N-(3-Oxobutanoyl)-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.

Formula: C₈H₁₁NO₄
MW: 185.2 g/mol
Storage Temperature: Freezer
CAS Number: 106983-29-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color White solid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR