N-Octanoyl-L-homoserine lactone 98
Supplier: ADIPOGEN CORP MS
Synonyms:
(S)-N-(2-Oxotetrahydrofuran-3-yl)octanamide, N-[(3S)-Tetrahydro-2-oxo-3-furanyl]-octanamide
CDX-O0038-M005
CDX-O0038-M050
102990-458EA
88.62
USD
102990-458
102990-460
N-Octanoyl-L-homoserine lactone 98
N-Octanoyl-L-homoserine lactone
N-Octanoyl-L-homoserine lactone (C8-HSL) is a small diffusible signaling molecule involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. The applications of this molecule include infection prevention and regulation of virulence in general and in cystic fibrosis.
Formula:
C₁₂H₂₁NO₃ MW: 227.3 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11113139 CAS Number: 147852-84-4 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in DMSO and dimethylformamide (approx. 30mg/ml). |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...