N-(3-Oxobutanoyl)-L-homoserine lactone ≥97% (by GC)
Supplier: ADIPOGEN CORP MS
Synonyms:
3-oxo-C4-HSL
CDX-O0037-M010
CDX-O0037-M100
102990-454EA
263.08
USD
102990-454
102990-456
N-(3-Oxobutanoyl)-L-homoserine lactone ≥97% (by GC)
N-(3-Oxobutanoyl)-L-homoserine lactone
N-(3-Oxobutanoyl)-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Formula:
C₈H₁₁NO₄ MW: 185.2 g/mol Storage Temperature: Freezer |
CAS Number:
148433-27-6 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...