N-(3-Oxododecanoyl)-L-homoserine lactone ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
3-Oxo-N-[(3S)-2-oxotetrahydro-3-furanyl]dodecanamide
CDX-O0031-M010
CDX-O0031-M020
CDX-O0031-M100
102990-682EA
88.62
USD
102990-682
102990-684
102990-686
N-(3-Oxododecanoyl)-L-homoserine lactone ≥98% (by NMR)
N-(3-Oxododecanoyl)-L-homoserine lactone
Quorum sensing agent.
Formula:
C₁₆H₂₇NO₄ MW: 297.39 g/mol Storage Temperature: Freezer |
CAS Number:
168982-69-2 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...