N-Octadecanoyl-Nile Blue ≥98% (by TLC)

Supplier: ADIPOGEN CORP MS

Synonyms: Chromoionophore I, ETH 5294, 3-Octadecanoylimino-7-(diethylamino)-1,2-benzophenoxazine,

CDX-O0027-M010 CDX-O0027-M020 CDX-O0027-M100
102990-676EA 121.85 USD
102990-676 102990-678 102990-680
N-Octadecanoyl-Nile Blue ≥98% (by TLC)
N-Octadecanoyl nile blue

This membrane probe has been used as an optode sensor for sodium and other ions. It was used to set up optical dihydrogen phosphate-selective sensors better suited for operation at physiological pH than their ion selective electrode counterparts.

Formula: C₃₈H₅₃N₃O₂
MW: 583.85 g/mol
Storage Temperature: Freezer
CAS Number: 125829-24-5

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (TLC)
Appearance/Color Solid.
Solubility Soluble in alcohols, DMSO or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR