4-Methylumbelliferyl heptanoate ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

CDX-O0007-M100 CDX-O0007-M500
102990-658EA 55.38 USD
102990-658 102990-660
4-Methylumbelliferyl heptanoate ≥98% (by HPLC)
4-Methylumbelliferyl heptanoate

Fluorogenic substrate for lipases.

Formula: C₁₇H₂₀O₄
Storage Temperature: Freezer
CAS Number: 18319-92-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Solid.
Solubility Soluble in alcohols or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR