Nordihydrocapsiate ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: 4-Hydroxy-3-methoxybenzyl 7-methyloctanoate, NDCT

CDX-N0121-M025 CDX-N0121-M100
102990-650EA 139.85 USD
102990-650 102990-652
Nordihydrocapsiate ≥95% (by HPLC)
Nordihydrocapsiate

Major capsaicinoid. TRPV1 and hTRPA1 agonist. Shown to activate capsaicin receptor, also called transient receptor potential vanilloid subfamily member 1 (TRPV1), but without inducing a nociceptive response. Anti-inflammatory and anticancer compound.

Formula: C₁₇H₂₆O₄
MW: 294.39 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28137841
CAS Number: 220012-53-3

Order Now

Specification Test Results

Source/Host Synthetic
Purity 95% (HPLC)
Appearance/Color Dark brown liquid .
Solubility Soluble in methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR