(2R)-3-(1-Naphthalenyloxy)-1,2-propanediol ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-N0073-M010
CDX-N0073-M025
102990-642EA
333.69
USD
102990-642
102990-644
(2R)-3-(1-Naphthalenyloxy)-1,2-propanediol ≥97% (by HPLC)
(2R)-3-(1-Naphthalenyloxy)-1,2-propanediol
Active metabolite of the anticonvulsant compound propranolol. Compound can be used as analytical reference material.
Formula:
C₁₃H₁₄O₃ Storage Temperature: Freezer |
MDL Number:
MFCD00673941 CAS Number: 61248-78-0 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in DMSO or methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...