Novaluron ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
1-[3-Chloro-4-(1,1,2-trifluoro-2-trifluoromethoxyethoxy)phenyl]-3-(2,6-difluorobenzoyl)urea
CDX-N0069-M100
CDX-N0069-M500
102990-638EA
220.15
USD
102990-638
102990-640
Novaluron ≥98% (by HPLC)
Novaluron
Acylurea insecticide. Insect growth inhibitor. Chitin synthase inhibitor active against larvae of Coleoptera, Homoptera, Diptera and Lepidoptera. Shown to be an inhibitor of acetylcholinesterase in B-biotype Bemisia tabaci. Compound can be used as analytical reference material.
Formula:
C₁₇H₉ClF₈N₂O₄ MW: 492.7 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD04112632 CAS Number: 116714-46-6 UN: 3077 ADR: 9,III |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White to off-white powder. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...