2-(2-Naphthyl)-2-(phenylmethoxy)-ethanenitrile ≥95% (by NMR)
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-N0052-M005
CDX-N0052-M025
CDX-N0052-M250
102990-620EA
106.62
USD
102990-620
102990-622
102990-624
2-(2-Naphthyl)-2-(phenylmethoxy)-ethanenitrile ≥95% (by NMR)
2-(2-Naphthyl)-2-(phenylmethoxy)ethanenitrile
This decarboxylated S-Adenosyl-L-methionine is an important metabolite in polyamine biosynthesis, acting as aminopropyl group donor for propylamine transferases such as spermine synthase and spermidine synthase.
Formula:
C₁₉H₁₅NO Storage Temperature: Freezer |
CAS Number:
500372-25-8 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...