NBD-undecanoic acid ≥98% (by TLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
11-(7-Nitrobenzofurazan-4-ylamino)undecanoic acid
CDX-N0011-M100
CDX-N0011-G001
102990-600EA
170.31
USD
102990-600
102990-602
NBD-undecanoic acid ≥98% (by TLC)
NBD-undecanoic acid
Fluorescent reagent with high selectivity.
Formula:
C₁₇H₂₄N₄O₅ MW: 364.4 g/mol Storage Temperature: Freezer |
CAS Number:
351002-77-2 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Dark red powder. |
Solubility | Soluble in methanol or DMF. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...