NBD-X ≥96% (by HPLC)

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: 6-(N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)amino)hexanoic acid, 6-(7-Nitrobenzofurazan-4-ylamino)-hexanoic acid

CDX-N0005-M200 CDX-N0005-G001
102990-582EA 236.77 USD
102990-582 102990-584
NBD-X ≥96% (by HPLC)
NBD-X

Fluorescent probe used for investigation of binding sites of fatty acid and sterol carrier proteins, also used for cell membrane staining.

Formula: C₁₂H₁₄N₄O₅
MW: 294.26 g/mol
Storage Temperature: Refrigerator
CAS Number: 88235-25-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 96% (HPLC)
Appearance/Color Orange-brown powder.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR