(S)-alpha-Methoxy-2-naphthylacetic acid ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Danger
|
CDX-M0213-M100
CDX-M0213-M500
102990-578EA
105.23
USD
102990-578
102990-580
(S)-alpha-Methoxy-2-naphthylacetic acid ≥97% (by HPLC)
(S)-alpha-Methoxy-2-naphthylacetic acid
Excellent chiral anisotropic NMR reagent. Used for the determination of the absolute configuration of secondary alcohols.
Formula:
C₁₃H₁₂O₃ Storage Temperature: Freezer |
MDL Number:
MFCD10000872 CAS Number: 157134-51-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...