(E)-8-Methyl-6-nonenoic acid ≥85%

Supplier: ADIPOGEN CORP MS

Synonyms: 8-Methylnonenoate

CDX-M0209-M500 CDX-M0209-G001
102990-574EA 519.23 USD
102990-574 102990-576
(E)-8-Methyl-6-nonenoic acid ≥85%
(E)-8-Methyl-6-nonenoic acid

Intermediate of the capsaicin biosynthesis. Fatty acid derivative from leucine/valine pathway. Plays a crucial role in determining the efficacy of capsaicin levels. The quantity of (E)-8-Methyl-6-nonenoic acid determines the pungency in placental tissues of Capsicum.

Formula: C₁₀H₁₈O₂
Storage Temperature: Freezer
MDL Number: MFCD00203855
CAS Number: 59320-77-3

Order Now

Specification Test Results

Source/Host Synthetic
Purity 85% (GC)
Appearance/Color Light yellow liquid.
Solubility Soluble in chloroform, dichloromethane or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR