N-Methylbis[(trifluoromethyl)sulfonyl]imide ≥90% (by GC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 1,1,1-Trifluoro-N-methyl-N-[(trifluoromethyl)sulphonyl]methanesulphonamide

CDX-M0154-GG25 CDX-M0154-G005 CDX-M0154-G025
102990-562EA 113.54 USD
102990-562 102990-564 102990-566
N-Methylbis[(trifluoromethyl)sulfonyl]imide ≥90% (by GC)
N-Methylbis[(trifluoromethyl)sulfonyl]imide

Powerful alkylation agent. Used for direct methylation and trifluoroethylation of imidazole and pyridine derivatives to produce a variety of room temperature ionic liquids (RTILs).

Formula: C₃H₃F₆NO₄S₂
MW: 295.18 g/mol
Storage Temperature: Freezer
MDL Number: MFCD23704393
CAS Number: 537031-78-0

Order Now

Specification Test Results

Source/Host Synthetic
Purity >90% (GC)
Appearance/Color Light green, light yellow liquid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR