Mesuximide ≥98% (by GC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Methsuximide

CDX-M0153-M050 CDX-M0153-M100
102990-558EA 210.46 USD
102990-558 102990-560
Mesuximide ≥98% (by GC)
Mesuximide

A succinimide with anti-epileptic and anti-convulsant properties. The mechanisms of action are unknown and possibly multiple. Metabolically reduced to it's pharmacologically active metabolite N-desmethylmethosuximide.

Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
Storage Temperature: Freezer
CAS Number: 77-41-8

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (GC)
Appearance/Color Yellow powder.
Solubility Soluble in chloroform or methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR