1-Methyltryptamine ≥95% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

CDX-M0138-M200 CDX-M0138-G001
102990-550EA 275.54 USD
102990-550 102990-552
1-Methyltryptamine ≥95% (by NMR)
1-Methyltryptamine

Deacetylase inhibitor.

Formula: C₁₁H₁₄N₂
Storage Temperature: Freezer
CAS Number: 7518-21-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (NMR)
Appearance/Color Solid.
Solubility Soluble in DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR